Merck
  • Ag2S-ZnO--an efficient photocatalyst for the mineralization of Acid Black 1 with UV light.
Ag2S-ZnO--an efficient photocatalyst for the mineralization of Acid Black 1 with UV light.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-16)
B Subash, B Krishnakumar, M Swaminathan, M Shanthi
ABSTRACT
The Ag(2)S loaded ZnO (Ag(2)S-ZnO) was successfully synthesized by precipitation of zinc oxalate and Ag(2)S and calcination of the mixed precipitate at 400 °C for 12 h. The catalyst was characterized by X-ray diffraction (XRD), scanning electron microscope (SEM) images, energy dispersive spectra (EDS), diffuse reflectance spectra (DRS) photoluminescence spectra (PL) and BET surface area measurements. The photocatalytic activity of Ag(2)S-ZnO was investigated for the degradation of Acid Black (AB 1) in aqueous solution using UV light. Ag(2)S-ZnO is found to be more efficient than commercial ZnO, prepared ZnO and TiO(2)-P25 at pH 9 for the mineralization of Acid Black 1. The effects of operational parameters such as the amount of photocatalyst, dye concentration, initial pH on photo mineralization have been analyzed. Mechanism of degradation by Ag(2)S-ZnO is proposed. The mineralization of Acid Black 1 has also been confirmed by COD measurements. The catalyst is found to be reusable.
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent