Merck
  • Use of specific neuromodulators in the treatment of chronic, idiopathic cough: a systematic review.
Use of specific neuromodulators in the treatment of chronic, idiopathic cough: a systematic review.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2013-01-10)
Seth M Cohen, Stephanie Misono
ABSTRACT
The goal of this systematic review was to examine the evidence for the use of the neuromodulating agents, amitriptyline, gabapentin, pregabalin, and baclofen, in the management of chronic, idiopathic cough patients. Online databases, including PubMed, Embase, Cochrane Review, and Web of Science, and publications cited in bibliographies were used. Literature was searched by the 2 authors with a priori criteria for study selection. Eight relevant articles were identified, including 2 randomized controlled trials, 2 prospective cohort or case-series designs with consecutive patients, 1 retrospective case series of consecutive patients, 1 retrospective case series whose consecutive status was not known, and 2 case reports of 6 and 2 patients, respectively. Improvements in cough-specific quality of life were noted in the randomized controlled trials. Cough severity was reduced in studies that measured this outcome measure. In the remaining studies, cough symptoms were less after neuromodulator treatment. Benefit from neuromodulator treatment with amitriptyline, gabapentin, pregabalin, and baclofen in chronic, idiopathic cough patients was demonstrated. Further investigations using objective and subjective outcome measures are needed as well as studies exploring optimal dose, length of treatment, and relapse rates posttreatment.
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Supelco
Amitriptyline hydrochloride solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-Baclofen, ≥98% (HPLC), solid