Merck
  • Structural and functional studies on a proline-rich peptide isolated from swine saliva endowed with antifungal activity towards Cryptococcus neoformans.
Structural and functional studies on a proline-rich peptide isolated from swine saliva endowed with antifungal activity towards Cryptococcus neoformans.
Biochimica et biophysica acta (2013-01-01)
Stefania Conti, Giorgia Radicioni, Tecla Ciociola, Renato Longhi, Luciano Polonelli, Rita Gatti, Tiziana Cabras, Irene Messana, Massimo Castagnola, Alberto Vitali
ABSTRACT
A proline-rich peptide of 2733Da, isolated from pig parotid granule preparations was tested against different pathogenic fungi. It showed interesting antifungal activity towards a clinical isolate of Cryptococcus neoformans, with an EC(50) of 2.2μM. Neither cytotoxic nor haemolytic effects were observed towards mammalian cells. Circular dichroism and infrared spectroscopic studies showed that the peptide adopted a combination of polyproline type-II, β-turn and unordered conformations at physiological temperatures. Temperature dependent experiments evidenced a tendency to adopt a polyproline-II helix conformation. From experiments with lipid vesicles, Neutral Red Uptake (NRU), haemolytic assays, and confocal microscopy studies, it could be hypothesized that the peptide may exert its antifungal effect by interacting with an intracellular target rather than through membrane damage.
Sigma-Aldrich
Poly-L-proline, mol wt >30,000
Sigma-Aldrich
Poly-L-proline, mol wt 1,000-10,000