Merck
  • Specific stress-induced storage of trehalose, glycerol and D-arabitol in response to oxidative and osmotic stress in Candida albicans.
Specific stress-induced storage of trehalose, glycerol and D-arabitol in response to oxidative and osmotic stress in Candida albicans.
Biochemical and biophysical research communications (2012-12-25)
Ruth Sánchez-Fresneda, José P Guirao-Abad, Alejandra Argüelles, Pilar González-Párraga, Eulogio Valentín, Juan-Carlos Argüelles
ABSTRACT
Candida albicans exponential yeast cells are able to face environmental challenges by mounting a rapid and efficient "general stress response". Here we show that one of the main components of this response consists of the intracellular protective accumulation of the non-reducing disaccharide trehalose and two polyols, glycerol and D-arabitol, an accumulation that occurs in a stress-specific dependent manner. Thus, oxidative exposures promoted a marked increase in both trehalose and D-arabitol in the wild type strain, RM-100, whereas the glycerol content remained virtually unaffected with respect to basal levels. In contrast, osmotic challenges induced the significant storage of glycerol accompanied by minor changes, or even a slight drop, in the intracellular content of trehalose and D-arabitol. We examined the hypothetical role in this process of the MAP kinase Hog1, which regulates the protective responses in C. albicans against both oxidative and osmotic stress. Interestingly, unlike glycerol synthesis, the stress-induced trehalose accumulation was always Hog1-independent, whereas the ability to synthesize D-arabitol was only partially dependent on a functional Hog1 pathway.
Sigma-Aldrich
D-(+)-Arabitol, ≥99% (GC)
L-Arabinitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-(−)-Arabitol, ≥98% (GC)