Merck
  • Inhibitory mechanism of pancreatic amyloid fibril formation: formation of the complex between tea catechins and the fragment of residues 22-27.
Inhibitory mechanism of pancreatic amyloid fibril formation: formation of the complex between tea catechins and the fragment of residues 22-27.
Biochemistry (2012-12-05)
Miya Kamihira-Ishijima, Hiromi Nakazawa, Atsushi Kira, Akira Naito, Tsutomu Nakayama
ABSTRACT
Islet amyloid polypeptide (IAPP) is a major component of pancreatic amyloid deposits associated with type 2 diabetes. Polyphenols contained in plant foods have been found to inhibit amyloid fibril formation of proteins and/or peptides. However, the inhibition mechanism is not clear for a variety of systems. Here the inhibition mechanism of green tea polyphenols, catechins, on amyloid fibril formation of the IAPP fragment (IAPP22-27), which is of sufficient length for formation of β-sheet-containing amyloid fibrils, was investigated by means of kinetic analysis. A quartz crystal microbalance (QCM) determined that the association constants of gallate-type catechins [epicatechin 3-gallate (ECg) and epigallocatechin 3-gallate] for binding to IAPP22-27 immobilized on the gold plate in QCM were 1 order of magnitude larger than those of the free IAPP22-27 peptide, and also those of epicatechin and epigallocatechin. Kinetic analysis using a two-step autocatalytic reaction mechanism revealed that ECg significantly reduced the rate constants of the first nucleation step of amyloid fibril formation, while the rate of autocatalytic growth was less retarded. (1)H nuclear magnetic resonance studies clarified that a IAPP22-27/ECg complex clearly forms as viewed from the (1)H chemical shift changes and line broadening. Our study suggests that tea catechins specifically inhibit the early stages of amyloid fibril formation to form amyloid nuclei by interacting with the unstructured peptide and that this inhibition mechanism is of great therapeutic value because stabilization of the native state could delay the pathogenesis of amyloid diseases and also the toxicity of the small oligomer (protofibril) is reported to be greater than that of the mature fibril.
Product Description
Supelco
(−)-Gallocatechin, analytical standard
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Epigallocatechin, primary reference standard
Epicatechin gallate, primary reference standard
Supelco
(−)-Epigallocatechin, analytical standard