- Tetrandrine citrate eliminates imatinib-resistant chronic myeloid leukemia cells in vitro and in vivo by inhibiting Bcr-Abl/β-catenin axis.
Tetrandrine citrate eliminates imatinib-resistant chronic myeloid leukemia cells in vitro and in vivo by inhibiting Bcr-Abl/β-catenin axis.
To evaluate the effects of tetrandrine citrate, a novel tetrandrine salt with high water solubility, on the growth of imatinib (IM)-resistant chronic myeloid leukemia (CML) in vitro and in vivo, and reveal action molecular mechanisms. Cell viability in vitro was measured using methyl thiazolyl tetrazolium (MTT) assay. CML cell growth in vivo was assessed using a xenograft model in nude mice. Bcr-Abl and β-catenin protein levels were determined using Western blotting. Bcr-Abl messenger RNA (mRNA) was measured by reverse transcription polymerase chain reaction (RT-PCR). Flow cytometry (FCM) was used to determine cell cycle status. Tetrandrine citrate inhibited the growth of IM-resistant K562 cells, primary leukemia cells, and primitive CD34(+) leukemia cells, and their inhibition concentration that inhibited 50% of target cells (IC(50)) ranged from 1.20 to 2.97 μg/ml. In contrast, tetrandrine citrate did not affect normal blood cells under the same conditions, and IC(50) values were about 10.12-13.11 μg/ml. Oral administration of tetrandrine citrate caused complete regression of IM-resistant K562 xenografts in nude mice without overt toxicity. Western blot results revealed that treatment of IM-resistant K562 cells with tetrandrine citrate resulted in a significant decrease of both p210(Bcr-Abl) and β-catenin proteins, but IM did not affect the Bcr-Abl protein levels. Proteasome inhibitor, MG132, did not prevent tetrandrine-mediated decrease of the p210(Bcr-Abl) protein. RT-PCR results showed that tetrandrine treatment caused a decrease of Bcr-Abl mRNA. FCM analysis indicated that tetrandrine induced gap 1 (G(1)) arrest in CML cells. Tetrandrine citrate is a novel orally active tetrandrine salt with potent anti-tumor activity against IM-resistant K562 cells and CML cells. Tetrandrine citrate-induced growth inhibition of leukemia cells may be involved in the depletion of p210(Bcr-Abl) mRNA and β-catenin protein.