Merck
  • Withaferin A suppresses tumor promoter 12-O-tetradecanoylphorbol 13-acetate-induced decreases in isocitrate dehydrogenase 1 activity and mitochondrial function in skin epidermal JB6 cells.
Withaferin A suppresses tumor promoter 12-O-tetradecanoylphorbol 13-acetate-induced decreases in isocitrate dehydrogenase 1 activity and mitochondrial function in skin epidermal JB6 cells.
Cancer science (2012-10-31)
Wenjuan Li, Yunfeng Zhao
ABSTRACT
Withaferin A (WA) is a bioactive compound derived from Withania somnifera. The antitumor activity of WA has been well studied in human cancer models; however, its chemopreventive potential is unclear. In the present study, we used the skin epidermal JB6 P+ cells, a well-established model for tumor promotion, and demonstrated that WA suppressed the tumor promoter 12-O-tetradecanoylphorbol 13-acetate (TPA)-induced cell transformation and cell proliferation. Interestingly, TPA inactivated isocitrate dehydrogenase 1 (IDH1), which was reversed by WA. Similar results were also observed in mouse skin tissue. Therefore, we focused on metabolism as the potential mechanism of action. We found that mitochondrial functions were downregulated by TPA treatment, as indicated by reduced mitochondrial membrane potential, complex I activity and mitochondrial respiration. However, all of these downregulations were inhibited by WA. In addition, we examined the levels of α-ketoglutarate, a product of IDH1, and WA blocked its reduction upon TPA treatment. Finally, we detected the lactate level as a glycolysis marker, and WA suppressed its elevation caused by tumor promoter treatment. Altogether, these results suggest that WA might exert its chemopreventive activity via inhibiting not only oncogenic activation, but also IDH1 inactivation and mitochondrial dysfunction in early tumorigenesis.
Sigma-Aldrich
Withaferin A, ≥95% (HPLC)
Supelco
Withaferin A, analytical standard
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, BioUltra
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, ≥98% (titration)
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Supelco
α-Ketoglutaric acid disodium salt hydrate, ≥95%
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)