Merck
  • Identification and characterization of novel catalytic bioscavengers of organophosphorus nerve agents.
Identification and characterization of novel catalytic bioscavengers of organophosphorus nerve agents.
Chemico-biological interactions (2012-10-09)
Tamara C Otto, Jennifer R Scott, Monika A Kauffman, Sean M Hodgins, Robert C Ditargiani, James H Hughes, Erin P Sarricks, Greg A Saturday, Tracey A Hamilton, Douglas M Cerasoli
ABSTRACT
In an effort to discover novel catalytic bioscavengers of organophosphorus (OP) nerve agents, cell lysates from a diverse set of bacterial strains were screened for their capacity to hydrolyze the OP nerve agents VX, VR, and soman (GD). The library of bacterial strains was identified using both random and rational approaches. Specifically, two representative strains from eight categories of extremophiles were chosen at random. For the rational approach, the protein sequence of organophosphorus hydrolase (OPH) from Brevundimonas diminuta was searched against a non-redundant protein database using the Basic Local Alignment Search Tool to find regions of local similarity between sequences. Over 15 protein sequences with significant sequence similarity to OPH were identified from a variety of bacterial strains. Some of these matches were based on predicted protein structures derived from bacterial genome sequences rather than from bona fide proteins isolated from bacteria. Of the 25 strains selected for nerve agent testing, three bacterial strains had measurable levels of OP hydrolase activity. These strains are Ammoniphilus oxalaticus, Haloarcula sp., and Micromonospora aurantiaca. Lysates from A. oxalaticus had detectable hydrolysis of VR; Haloarcula sp. had appreciable hydrolysis of VX and VR, whereas lysates from M. aurantiaca had detectable hydrolysis of VR and GD.
Product Description
Supelco
Paraoxon-ethyl, PESTANAL®, analytical standard
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil