Merck
  • Adsorption of poly(vinylformamide-co-vinylamine) polymers (PVFA-co-PVAm) on copper.
Adsorption of poly(vinylformamide-co-vinylamine) polymers (PVFA-co-PVAm) on copper.
Langmuir : the ACS journal of surfaces and colloids (2012-10-06)
Susan Seifert, Susanne Höhne, Frank Simon, Christian Hanzelmann, René Winkler, Thomas Schmidt, Ralf Frenzel, Petra Uhlmann, Stefan Spange
ABSTRACT
The adsorption of poly(vinylformamide) (PVFA) and its derivative statistical copolymer poly(vinyl-formamide-co-vinylamine) (PVFA-co-PVAm) on metallic copper and copper oxide particles as well as planar copper surfaces was studied as a function of the degree of hydrolysis of PVFA, the pH, and the polymer concentration in solution. The chemical composition and molecular structure of the PVFA-co-PVAm layers were investigated by surface-sensitive spectroscopic methods such as XPS, DRIFT spectroscopy, and ellipsometry. The findings allowed us to explain the adsorption mechanisms and the forces driving the PVFA-co-PVAm adsorption. It was shown that PVFA-co-PVAm layers thicker than 30 nm are able to protect the planar copper surface against corrosive attack.
Sigma-Aldrich
Copper(I) oxide, ≥99.99% trace metals basis, anhydrous