Merck
[A man with progressive diplopia].
Nederlands tijdschrift voor geneeskunde (2012-09-14)
Niels Crama, B Jeroen Klevering
ABSTRACT
A 71-year-old man presented with progressive diplopia due to swelling of a Miragel explant. Removal of the explant resulted in relief of symptoms.