Merck
  • Encapsulation of photosensitizer into multilayer microcapsules by combination of spontaneous deposition and heat-induced shrinkage for photodynamic therapy.
Encapsulation of photosensitizer into multilayer microcapsules by combination of spontaneous deposition and heat-induced shrinkage for photodynamic therapy.
Macromolecular bioscience (2012-09-12)
Yuanyuan Han, Jing Bu, Yuying Zhang, Weijun Tong, Changyou Gao
ABSTRACT
Annealing of PDADMAC/PSS multilayer microcapsules assembled on PSS-doped CaCO(3) particles at 80 °C for 30 min reduces their size dramatically from 6.9 ± 0.3 to 3.1 ± 0.5 µm. Methylene blue molecules are encapsulated by spontaneous deposition and post-annealing with a concentration of 22 mg · mL(-1), which is 1000 times higher than the feeding value. The unreleased MB molecules are retained stably for a long time, which are then protected by the capsules against reductive enzymes and keep their photodynamic activity. The viability of HeLa cells incubated with the MB-loaded capsules decreases sharply from ≈ 75 (dark cytotoxicity) to ≈ 20% after irradiation with a laser at 671 nm and 60 J · cm(-2) for 75 s.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(4-styrenesulfonic acid) solution, Mw ~75,000, 18 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~70,000
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~1,000,000, powder
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~200,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~1,000,000, 25 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~70,000, 30 wt. % in H2O