Merck
  • Enantioselective synthesis of anti- and syn-homopropargyl alcohols via chiral Brønsted acid catalyzed asymmetric allenylboration reactions.
Enantioselective synthesis of anti- and syn-homopropargyl alcohols via chiral Brønsted acid catalyzed asymmetric allenylboration reactions.
Journal of the American Chemical Society (2012-06-27)
Ming Chen, William R Roush
ABSTRACT
Chiral Brønsted acid catalyzed asymmetric allenylboration reactions are described. Under optimized conditions, anti-homopropargyl alcohols 2 are obtained in high yields with excellent diastereo- and enantioselectivities from stereochemically matched aldehyde allenylboration reactions with (M)-1 catalyzed by the chiral phosphoric acid (S)-4. The syn-isomers 3 can also be obtained in good diastereoselectivities and excellent enantioselectivities from the mismatched allenylboration reactions of aromatic aldehydes using (M)-1 in the presence of the enantiomeric phosphoric acid (R)-4. The stereochemistry of the methyl group introduced into 2 and 3 is controlled by the chirality of the allenylboronate (M)-1, whereas the configuration of the new hydroxyl stereocenter is controlled by the enantioselectivity of the chiral phosphoric acid catalyst used in these reactions. The synthetic utility of this methodology was further demonstrated in highly diastereoselective syntheses of a variety of anti, anti-stereotriads, the direct synthesis of which has constituted a significant challenge using previous generations of aldol and crotylmetal reagents.
Sigma-Aldrich
Propargyl alcohol, 99%
Pricing and availability is not currently available.