Merck
Ultrasound-triggered thrombolysis using urokinase-loaded nanogels.
International journal of pharmaceutics (2012-06-12)
Haiqiang Jin, Hui Tan, Lingling Zhao, Weiping Sun, Lijun Zhu, Yongan Sun, Hongjun Hao, Haiying Xing, Linlin Liu, Xiaozhong Qu, Yining Huang, Zhenzhong Yang
ABSTRACT
To find a way to modulate the effect of thrombolytic proteins by increasing their specificity, minimizing their adverse effect as well as lengthening their circulation time for the treatment of ischemic vascular disease holds great promise. In this work, urokinase-type plasminogen activator (uPA) was encapsulated into hollow nanogels which are generated by the reaction of glycol chitosan and aldehyde capped poly(ethylene glycol) (OHC-PEG-CHO) through a one-step approach of ultrasonic spray. The uPA-loaded nanogels, with size of 200-300 nm, have longer circulation time than that of the nude urokinase in vivo, besides the protein can be triggered to release in faster rate under diagnostic ultrasonic condition of 2 MHz, which significantly enhanced the thrombolysis of clots. The results are promising for increasing the specificity and positive effects of thrombolytic agents like recombinant tissue plasminogen activator (rt-PA) for the current treatment of ischemic vascular disease.
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Supelco
Benzaldehyde, analytical standard
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material