Merck
  • [Effect of polysaccharides on biological activity of human lactoferrin].
[Effect of polysaccharides on biological activity of human lactoferrin].
Prikladnaia biokhimiia i mikrobiologiia (2012-02-01)
N M Mestechkina, O A Bezborodova, A V Il'ina, A N Levov, S Iu Kleĭmenov, E R Nemtsova, R I Iakubovskaia, V D Shcherbukhin, V P Varlamov
ABSTRACT
The influence of neutral and ionic polysaccharides on the antioxidant (AOA) and detoxifying activities of lactoferrin (LF) and the duration of its circulation in the body was studied. In addition to natural polymers, we studied artificial chitosan derivatives with different functional groups. On the basis ofAOA test, five polysaccharides were selected. The study of the detoxifying effect of LF in two models of induced toxicity revealed polysaccharides that maintained or increased the detoxifying activity of LF. We established that the formation of a complex of lactoferrin with two galactomannans and succinyl chitosan caused positive changes in LF properties: the detoxifying activity of the protein remained unchanged or increased, whereas its elimination from the body was decelerated.
Sigma-Aldrich
(+)-Arabinogalactan, ≥80% (HPLC), from larch wood
Pricing and availability is not currently available.