Merck
  • Highly stable and efficient Ag/AgCl@TiO2 photocatalyst: preparation, characterization, and application in the treatment of aqueous hazardous pollutants.
Highly stable and efficient Ag/AgCl@TiO2 photocatalyst: preparation, characterization, and application in the treatment of aqueous hazardous pollutants.
Journal of hazardous materials (2011-12-20)
Jian-Feng Guo, Bowen Ma, Anyuan Yin, Kangnian Fan, Wei-Lin Dai
ABSTRACT
A new plasmonic photocatalyst of Ag-AgCl@TiO(2) was prepared by deposition-precipitation and photoreduction. This photocatalyst exhibited efficient photocatalytic activity for the degradation of 4-chlorophenol and photoreduction of Cr(VI) ion under visible light irradiation. Its high photocatalytic activity can be attributed to the surface plasmon resonance effect of Ag nanoparticles, which were highly dispersed on the surface of Ag-AgCl@TiO(2). N(2) adsorption and desorption isotherm spectra, X-ray diffraction, X-ray photoelectron spectroscopy, and transmission electron microscopy were used to determine the correlation between the micro-structure and the catalytic properties of the as-prepared photocatalysts.
Sigma-Aldrich
Silver chloride, ReagentPlus®, 99%
Sigma-Aldrich
Silver chloride, 99.999% trace metals basis
Sigma-Aldrich
Silver chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis