Merck
  • Stimulation of the extracellular Ca²⁺-sensing receptor by denatonium.
Stimulation of the extracellular Ca²⁺-sensing receptor by denatonium.
Biochemical and biophysical research communications (2011-12-07)
Olga A Rogachevskaja, Gleb D Churbanov, Marina F Bystrova, Roman A Romanov, Stanislav S Kolesnikov
ABSTRACT
The extracellular Ca(2+)-sensing receptor (CASR) is a promiscuous G-protein-coupled receptor closely related to the taste receptors T1R1-T1R3. Here we analyzed the possibility that apart from being stimulated by external Ca(2+) and amino acids, the substances effective as tastants, CASR might serve as a receptor for other sapid compounds. CASR was heterologously expressed in HEK-293 cells, and their responsivity to a variety of bitter and sweet substances was examined. Among them, solely denatonium was found to stimulate Ca(2+) signaling in CASR-positive HEK-293 cells. Apparently, these Ca(2+) responses were specific, as those were inhibited by the CASR antagonist NSP-4123. Altogether, our findings indicate that denatonium stimulates CASR by shifting a dose-response curve for the principal CASR agonist Ca(2+) to lower concentrations.
Sigma-Aldrich
Sucrose octaacetate, ≥97%, FG
Sigma-Aldrich
D-(+)-Sucrose octaacetate, 98%
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
Supelco
Aspartame, analytical standard
Aspartame, European Pharmacopoeia (EP) Reference Standard