- Toxicity, sublethal effects, and performance of sulfluramid against the German cockroach (Dictyoptera: Blattellidae).
Toxicity, sublethal effects, and performance of sulfluramid against the German cockroach (Dictyoptera: Blattellidae).
Topical and oral toxicity of sulfluramid (N-ethyl perfluorooctane sulfonamide) were determined for the German cockroach, Blattella germanica (L.). The topical LD50 of sulfluramid was 175.6 micrograms/g for adult males, 117.8 micrograms/g for adult females, and 122.3 micrograms/g for gravid adult females. Ingestion increased toxicity approximately 1.4 times for adult male B. germanica. Twenty-four hours after topical treatment with 20 micrograms/insect sulfluramid, the percentage of female cockroaches that dropped their oothecae increased approximately 50% compared with controls treated with acetone. Sulfluramid also decreased oothecal hatch of both dropped and retained oothecae. Approximately 90% of oothecae from untreated females hatches, whereas less than 20% hatched from females treated with 20 micrograms/insect. Mean time for oothecal hatch increased linearly with increasing sulfluramid concentration. In arena studies in Ebeling choice boxes, LT50's ranged between approximately 2.3 and 3.9 d for a 0.331 mg/cm2 deposit and a 1.5% bait, respectively. Higher concentrations of sulfluramid were more repellent in both bait and residual formulations. Performance index values indicated excellent potential field efficacy. Field trials with 1.0 and 1.5% (AI) baits showed up to a 71.3% reduction in cockroach numbers. Baits controlled cockroaches throughout the 12-wk test.