Merck
  • Synthesis of Au nanoparticles decorated graphene oxide nanosheets: noncovalent functionalization by TWEEN 20 in situ reduction of aqueous chloroaurate ions for hydrazine detection and catalytic reduction of 4-nitrophenol.
Synthesis of Au nanoparticles decorated graphene oxide nanosheets: noncovalent functionalization by TWEEN 20 in situ reduction of aqueous chloroaurate ions for hydrazine detection and catalytic reduction of 4-nitrophenol.
Journal of hazardous materials (2011-10-25)
Wenbo Lu, Rui Ning, Xiaoyun Qin, Yingwei Zhang, Guohui Chang, Sen Liu, Yonglan Luo, Xuping Sun
ABSTRACT
In this paper, we develop a cost-effective and simple route for the synthesis of Au nanoparticles (AuNPs) decorated graphene oxide (GO) nanosheets using polyoxyethylene sorbitol anhydride monolaurate (TWEEN 20) as a stabilizing agent for GO as well as a reducing and immobilizing agent for AuNPs. The AuNPs assemble on the surface of TWEEN-functionalized GO by the in situ reduction of HAuCl(4) aqueous solution. The morphologies of these composites were characterized by atomic force microscopy (AFM) and transmission electron microscopy (TEM). It is found that the resultant AuNPs decorated GO nanosheets (AuNPs/TWEEN/GO) exhibit remarkable catalytic performance for hydrazine oxidation. This hydrazine sensor has a fast amperometric response time of less than 3s. The linear range is estimated to be from 5 μM to 3 mM (r=0.999), and the detection limit is estimated to be 78 nM at a signal-to-noise ratio of 3. The AuNPs/TWEEN/GO composites also exhibit good catalytic activity toward 4-nitrophenol (4-NP) reduction and the GO supports also enhance the catalytic activity via a synergistic effect.
Sigma-Aldrich
Sodium tetrachloroaurate(III) dihydrate, 99%
Sigma-Aldrich
Gold(III) chloride solution, 99.99% trace metals basis, 30 wt. % in dilute HCl
Sigma-Aldrich
TWEEN® 20, for synthesis
Sigma-Aldrich
Potassium gold(III) chloride, 99.995% trace metals basis
Sigma-Aldrich
Potassium gold(III) chloride, 98%