Merck
  • Evaluation of the indoxyl acetate hydrolysis test for the differentiation of Campylobacters.
Evaluation of the indoxyl acetate hydrolysis test for the differentiation of Campylobacters.
Journal of clinical microbiology (1990-06-01)
D S Hodge, A Borczyk, L L Wat
ABSTRACT
Indoxyl acetate hydrolysis is a rapid, inexpensive differential test which can be performed easily to help identify campylobacter. A total of 571 Campylobacter cultures, including atypical variants, representing 10 species was tested.
Sigma-Aldrich
Indoxyl acetate, ≥95% (GC)