Merck
  • Isolation and chromatographic analysis of bioactive triterpenoids from the bark extract of Cariniana domestica (Mart) Miers.
Isolation and chromatographic analysis of bioactive triterpenoids from the bark extract of Cariniana domestica (Mart) Miers.
Natural product research (2011-08-11)
Vanessa Janovik, Aline Augusti Boligon, Janaína Kieling Frohlich, Thiago Guilherme Schwanz, Tarciele Venturini Pozzebon, Sydney Hartz Alves, Margareth Linde Athayde
ABSTRACT
The Lecythidaceae family is composed of 25 genera and 400 species that occur in the form of trees with a pantropical distribution. The genera Cariniana belongs to the family Lecythidaceae, and there are few reports considering these species. In this work, fractionation of the dichloromethane fraction obtained from the aqueous extract of the stem bark of Cariniana domestica (Mart) Miers, popularly known as Jequitibá-roxo, led to the isolation of two mixtures of triterpenoids: lupeol and β-amyrin and β-sitosterol and stigmasterol. The structures of the isolated compounds were elucidated by spectroscopic (NMR) and chromatographic (GC-MS) techniques as well as literature data comparisons. β-Sitosterol, stigmasterol, lupeol and β-amyrin were quantified in dichloromethane fraction by high-performance liquid chromatography (HPLC/DAD). The dichloromethane fraction was also investigated for antioxidant and antifungal activities. The isolated compounds and their biological activities are reported for the first time for the species C. domestica.
Product Description
Supelco
β-Amyrin, analytical standard
Supelco
α-Amyrin, analytical standard