Merck
  • Metal-organic-framework-based tandem molecular sieves as a dual platform for selective microextraction and high-resolution gas chromatographic separation of n-alkanes in complex matrixes.
Metal-organic-framework-based tandem molecular sieves as a dual platform for selective microextraction and high-resolution gas chromatographic separation of n-alkanes in complex matrixes.
Analytical chemistry (2011-08-02)
Na Chang, Zhi-Yuan Gu, He-Fang Wang, Xiu-Ping Yan
ABSTRACT
Metal-organic frameworks (MOFs) were employed to design tandem molecular sieves as a dual platform for selective solid-phase microextraction (SPME) and high-resolution gas chromatographic (GC) separation of target analytes in complex matrixes. An elegant combination of a ZIF-8-coated fiber for SPME with a ZIF-8-coated capillary for GC allows selective extraction and separation of n-alkanes from complex matrixes such as petroleum-based fuel and biological fluids. The proposed tandem ZIF-8 molecular sieves not only offered good enhancement factors from 235 (hexane) to 1212 (nonane), but also exhibited wide linearity with 3 orders of magnitude for the tested linear alkanes. The limits of detection for the linear alkanes ranged from 0.46 ng L(-1) (nonane) to 1.06 ng L(-1)(hexane). The relative standard deviations of retention time, peak area, peak height, and half peak width for five replicate determinations of the tested n-alkanes at 30 ng L(-1) were 0.02-0.26%, 1.9-8.6%, 1.4-6.0%, and 1.3-7.2%, respectively. The developed tandem ZIF-8 molecular sieves were further used for the determination of linear alkanes in petroleum-based fuel and human serum. The large diversity in structure and pore size allows various combinations of MOFs for designing an MOF-based tandem molecular sieve platform to achieve different selectivities in extraction and chromatographic separation and to solve headache problems in complex real sample analysis.
Product Description
Supelco
Nonane, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nonane, anhydrous, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nonane, ReagentPlus®, 99%