Merck
  • Long-wavelength fluorimetric determination of food antioxidant capacity using Nile blue as reagent.
Long-wavelength fluorimetric determination of food antioxidant capacity using Nile blue as reagent.
Journal of agricultural and food chemistry (2011-03-04)
J Godoy-Navajas, M P Aguilar Caballos, A Gómez-Hens
ABSTRACT
A method for the determination of the antioxidant capacity using long-wavelength fluorescence measurements is described for the first time. This method is a modification of the conventional oxygen radical absorbance capacity (ORAC) method that uses fluorescein or phycoerythrin and the generator of peroxyl radicals, 2,2'-azo-bis-(2-methylpropionamidine) dihydrochloride (AAPH). The long-wavelength fluorophor nile blue is proposed as an analytical reagent alternative to these conventional fluorophores. Kinetic curves have been obtained by monitoring the fluorescence variation (λex, 620; λem, 680 nm) with time, using the 96-well microplate format. The vitamin E analogue 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid (Trolox) has been chosen as the model analyte, and the normalized area under the decay curve has been used as the analytical parameter. The dynamic range of the calibration curve is 0.8-8.0 μM, and the detection limit is 0.45 μM. The precision of the method, expressed as relative standard deviation and assayed using 1 and 5 μM Trolox concentrations, was 5.6 and 2.9%, respectively. The method has been applied to the analysis of fruit juices and wines, obtaining results that did not differ significantly from those provided using the ORAC method with fluorescein as reagent.
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %