Merck
  • Controlled photophysical behaviors between dibenzo-24-crown-8 bearing terpyridine moiety and fullerene-containing ammonium salt.
Controlled photophysical behaviors between dibenzo-24-crown-8 bearing terpyridine moiety and fullerene-containing ammonium salt.
The Journal of organic chemistry (2011-02-16)
Zhi-Jun Ding, Ying-Ming Zhang, Xue Teng, Yu Liu
ABSTRACT
A novel [2]pseudorotaxane was successfully constructed by the complexation of dibenzo[24]-crown-8 (DB24C8) derivative bearing terpyridine moiety (1) with lanthanide ion (Tb(3+)) and fullerene-containing ammonium salt (2), exhibiting the controlled photophysical behaviors as a reversible luminescent lanthanide switch in the presence of K(+) or 18-crown-6 (18C6).
Sigma-Aldrich
Dibenzo-24-crown-8, 98%