Merck
  • Strong suppression of the renin-angiotensin system has a renal-protective effect in hypertensive patients: high-dose ARB with ACE inhibitor (Hawaii) study.
Strong suppression of the renin-angiotensin system has a renal-protective effect in hypertensive patients: high-dose ARB with ACE inhibitor (Hawaii) study.
Hypertension research : official journal of the Japanese Society of Hypertension (2010-08-13)
Mitsuru Ohishi, Yasushi Takeya, Yuji Tatara, Koichi Yamamoto, Miyuki Onishi, Yoshihiro Maekawa, Kei Kamide, Hiromi Rakugi
ABSTRACT
The principal means for reducing proteinuria in patients with chronic kidney disease are strong blockade of the renin-angiotensin system and strict regulation of blood pressure (BP). This study compared the efficacy of the maximum permissible doses of two common angiotensin receptor blockers (ARBs), namely valsartan (maximum dose=160 mg per day) and olmesartan (maximum dose=40 mg per day). We also investigated whether a high-dose ARB or the combination of an angiotensin-converting enzyme inhibitor with a high-dose ARB would be more renal protective. We recruited 87 poorly controlled hypertensive patients. In the first study, 50 patients without proteinuria were switched from valsartan (160 mg per day) to olmesartan (40 mg per day) for 4 months. In the second study, 37 patients with proteinuria were randomized to either switch from valsartan 160 mg per day to 40 mg per day olmesartan (n=19; Olm-G) or addition of 2.5-10 mg per day imidapril (stepped up by 2.5 mg per month) to valsartan at 160 mg per day (n=18; Imi-G). After 4 months, the BP level decreased (first study) from 157/88 mm Hg to 145/82 mm Hg (P<0.001) and (second study) from 149/86 mm Hg to 135/77 mm Hg and 145/82 mm Hg for Olm-G and Imi-G, respectively. Furthermore, in the second study, urinary protein/creatinine excretion was reduced from 2.0±1.8 g g⁻¹ to 0.8±0.8 g g⁻¹ (P=0.0242) in Olm-G and from 1.4±1.3 g g⁻¹ to 0.9±1.0 g g⁻¹ (P=0.0398) in Imi-G. The significance persisted after adjustment for BP or other risk factors. Our results suggested that the maximum dose of olmesartan was more effective than that of valsartan and comparable with the combination of valsartan and imidapril for reducing BP and proteinuria in poorly controlled hypertensive patients.
Sigma-Aldrich
Imidapril hydrochloride, ≥98% (HPLC)