Merck
  • Intoxication by large amounts of barium nitrate overcome by early massive K supplementation and oral administration of magnesium sulphate.
Intoxication by large amounts of barium nitrate overcome by early massive K supplementation and oral administration of magnesium sulphate.
Human & experimental toxicology (2010-04-01)
Christine Payen, Arnaud Dellinger, Corine Pulce, Vincent Cirimele, Vincent Carbonnel, Pascal Kintz, Jacques Descotes
ABSTRACT
Suicide by ingestion of barium is exceptionally rare. Adverse health effects depend on the solubility of the barium compound. Severe hypokalemia, which generally occurs within 2 hours after ingestion, is the predominating feature of acute barium toxicity, subsequently leading to adverse effects on muscular activity and cardiac automaticity. We report one case of acute poisoning with barium nitrate, a soluble barium compound. A 75-year-old woman was hospitalized after suicidal ingestion of a burrow mole fumigant containing 12.375 g of barium nitrate. About 1 hour post-ingestion, she was only complaining of abdominal pain. The ECG recording demonstrated polymorphic ventricular premature complexes (VPCs). Laboratory data revealed profound hypokalemia (2.1 mmol/L). She made a complete and uneventful recovery after early and massive potassium supplementation combined with oral magnesium sulphate to prevent barium nitrate absorption.
Sigma-Aldrich
Barium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Barium nitrate, ACS reagent, ≥99%