MilliporeSigma
  • Gadolinium(III) complexes of 1,4,7-triazacyclononane based picolinate ligands: simultaneous optimization of water exchange kinetics and electronic relaxation.
Gadolinium(III) complexes of 1,4,7-triazacyclononane based picolinate ligands: simultaneous optimization of water exchange kinetics and electronic relaxation.
Dalton transactions (Cambridge, England : 2003) (2009-09-23)
Aline Nonat, Marion Giraud, Christelle Gateau, Pascal H Fries, Lothar Helm, Marinella Mazzanti
ABSTRACT
The two new tripodal picolinate H(3)ebpatcn (1-carboxyethyl-4,7-bis((6-carboxypyridin-2-yl)methyl)-1,4,7-triazacyclononane) and H(4)pbpatcn (1-methylphosphonic-acid-4,7-bis((6-carboxypyridin-2-yl)methyl)-1,4,7-triazacyclononane) ligands based on the 1,4,7-triazacyclononane anchor were prepared and their lanthanide complexes were characterized by NMR, fluorescence and potentiometric studies. The [Gd(ebpatcn)(H(2)O)] complex displays a relaxivity of r(1) = 4.68 mM(-1) s(-1) at 45 MHz and 298 K, whereas r(1) = 4.55 mM(-1) s(-1) was measured for [Gd(Hpbpatcn)(H(2)O)] under the same conditions. The modified scaffold of the ligands with respect to the previously reported H(3)bpatcn (1-(carboxymethyl)-4,7-bis[(6-carboxypyridin-2-yl)methyl]-1,4,7-triazacyclononane) leads to an optimization of the properties of these gadolinium complexes. The replacement of an acetate binding group of the H(3)bpatcn ligand with a propionate group (H(3)ebpatcn) or a phosphonate group (H(4)pbpatcn) leads to a faster exchange rate of the coordinated water molecule in both mono-aquo gadolinium complexes. The resulting water exchange rate is optimized for the future design of high relaxivity macromolecular gadolinium based contrast agents with a value measured by O(17) NMRD of k(ex) = 34 x 10(6) s(-1) for [Gd(Hpbpatcn)(H(2)O)] falling in the range of optimum values of (30 to 50) x 10(6) s(-1) predicted by the SBM theory. The water exchange rate k(ex)(298) = 86 x 10(6) s(-1) of the complex [Gd(ebpatcn)(H(2)O)] is the fastest reported in the literature for a neutral complex with only one inner-sphere water molecule. The relatively high stability of these modified gadolinium complexes (pGd = 14.1 for Gd(pbpatcn) and 13.1 for Gd(ebpatcn)) is similar to that of the [Gd(bpatcn)(H(2)O)] complex (pGd = 13.6). The high luminescence efficiency is also retained for the terbium complex. However, whereas the longitudinal electronic spin relaxation time keeps a value for [Gd(ebpatcn)(H(2)O)], which is long enough not to affect the relaxivity in macromolecular complexes (transient ZFS amplitude Delta(2) [10(20) rad(2) s(-2)] = 0.39), the O(17) relaxation and the (1)H NMRD indicate a rather fast electron spin relaxation for the phosphonate containing complex (Delta(2) [10(20) rad(2) s(-2)] = 1.3).
Sigma-Aldrich
1,4,7-Triazacyclononane, 95%
Pricing and availability is not currently available.