Merck
  • In vitro radioprotection studies of organoselenium compounds: differences between mono- and diselenides.
In vitro radioprotection studies of organoselenium compounds: differences between mono- and diselenides.
Radiation and environmental biophysics (2009-09-17)
B Santhosh Kumar, Amit Kunwar, A Ahmad, L B Kumbhare, V K Jain, K I Priyadarsini
ABSTRACT
Organoselenium compounds belonging to the class of monoselenides, such as selenomethionine (SeM) and methylselenocysteine (MSeCys) and diselenides including selenocystine (SeCys) and selenopropionic acid (SePA), were examined for their comparative radioprotective effects using in vitro models. Effects of these compounds on the inhibition of gamma-radiation induced lipid peroxidation in liposomes, protein carbonylation in bovine serum albumin (BSA) and strand breaks in pBR322 plasmid DNA, assessed, respectively, by the formation of thiobarbituric acid reactive substances, formation of 2,2'-dinitrophenyl hydrazine (DNPH) carbonyl complex and horizontal gel electrophoresis, were used to compare their radioprotective ability. The IC 50 values for SeCys, SePA, SeM and MSeCys for lipid peroxidation were 27 +/- 1, 33 +/- 2, 200 +/- 8 and 163 +/- 4 microM, respectively, and the values for inhibition of protein carbonylation were >200, 300 +/- 6, 464 +/- 8 and 436 +/- 3 microM, respectively. Inhibition of DNA strand break formation was tested at 200 microM for all the compounds and SePA and SeCys exhibited a protective effect on DNA, while SeM and MSeCys did not lead to any protection. The in vitro cytotoxicity studies in normal and tumor cells revealed that MSeCys and SeM were not cytotoxic to lymphocytes and EL4 tumor cells at the concentrations employed. In contrast, SeCys was toxic, with a higher effect on tumor cells than lymphocytes. Our studies suggest that the non-toxic diselenides like SePA should be explored as protective agents against gamma-irradiation induced damage.
Sigma-Aldrich
Seleno-L-cystine, 95%
Sigma-Aldrich
Seleno-DL-cystine, powder