Merck
  • Phospholipid decoration of microcapsules containing perfluorooctyl bromide used as ultrasound contrast agents.
Phospholipid decoration of microcapsules containing perfluorooctyl bromide used as ultrasound contrast agents.
Biomaterials (2008-12-23)
Raquel Díaz-López, Nicolas Tsapis, Danielle Libong, Pierre Chaminade, Carole Connan, Mohamed M Chehimi, Romain Berti, Nicolas Taulier, Wladimir Urbach, Valérie Nicolas, Elias Fattal
ABSTRACT
We present here an easy method to modify the surface chemistry of polymeric microcapsules of perfluorooctyl bromide used as ultrasound contrast agents (UCAs). Capsules were obtained by a solvent emulsification-evaporation process with phospholipids incorporated in the organic phase before emulsification. Several phospholipids were reviewed: fluorescent, pegylated and biotinylated phospholipids. The influence of phospholipid concentration on microcapsule size and morphology was evaluated. Only a fraction of the phospholipids is associated to microcapsules, the rest being dissolved with the surfactant in the aqueous phase. Microscopy shows that phospholipids are present within the shell and that the core/shell structure is preserved up to 0.5 mg fluorescent phospholipids, up to about 0.25 mg pegylated phospholipids or biotinylated phospholipids (for 100 mg of polymer, poly(lactide-co-glycolide) (PLGA)). HPLC allows quantifying phospholipids associated to capsules: they correspond to 10% of pegylated phospholipids introduced in the organic phase. The presence of pegylated lipids at the surface of capsules was confirmed by X-ray photon electron spectroscopy (XPS). The pegylation did not modify the echographic signal arising from capsules. Finally biotinylated microcapsules incubated with neutravidin tend to aggregate, which confirms the presence of biotin at the surface. These results are encouraging and future work will consist of nanocapsule surface modification for molecular imaging.
Sigma-Aldrich
1-Bromoheptadecafluorooctane, 99%
Avanti
DSPE-PEG(2000) Biotin, Avanti Research - A Croda Brand 880129C
Avanti
DSPE-PEG(2000) Biotin, Avanti Research - A Croda Brand 880129P, powder