Merck
  • [The mechanisms of 232Th and of Ce (III) toxic action on Chlorella vulgaris Beijer. Estimation of 232Th radiation impact contribution into toxic effect induced].
[The mechanisms of 232Th and of Ce (III) toxic action on Chlorella vulgaris Beijer. Estimation of 232Th radiation impact contribution into toxic effect induced].
Radiatsionnaia biologiia, radioecologiia (2008-08-12)
T I Evseeva, T A Maĭstrenko, S A Geras'kin, E S Belykh
ABSTRACT
232Th and Ce (III) toxic effects and its modifications with caffeine and D,L-buthionine-(S,R)-sulphoximine on Chlorella vulgaris Beijer were studied using an optical density measure after 24 hours growth. Concentrations of 232Th or of Ce--toxic effect relationship were shown to be nonlinear. In the first (nontoxic) concentration range (for Ce 0.036-1.642 micromol/L and for 232Th 0.001-1.551 micromol/L) algae biomass production registered by optical density do not significantly differ from the control one. In the second (toxic) concentration range dose-effect relationship for 232Th is characterized with quadratic dependence and in the case of Ce--with exponential dependence. 232Th radiation component contribution into effect observed is appeared as induction of DNA damages additional to spontaneous at the radionuclide concentration (equal to 0.345 micromol/L) that is three times lower than in case of the non-radioactive chemical analog Ce (1.071 micromol/L).
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.9% trace metals basis
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis