Merck
  • Synthesis of 1-/2-substituted-[1,2,3]triazolo[4,5-g]phthalazine-4,9-diones and evaluation of their cytotoxicity and topoisomerase II inhibition.
Synthesis of 1-/2-substituted-[1,2,3]triazolo[4,5-g]phthalazine-4,9-diones and evaluation of their cytotoxicity and topoisomerase II inhibition.
Bioorganic & medicinal chemistry (2008-03-07)
Jin Sung Kim, Hee-Kyung Rhee, Hyen Joo Park, Sang Kook Lee, Chong-Ock Lee, Hea-Young Park Choo
ABSTRACT
Studies on antitumor heterocyclic quinones containing nitrogens revealed that the number and position of nitrogens on the heterocyclic ring have significance on cytotoxicity of quinones. In our continuous effort to find more cytotoxic quinone compounds, we designed triazolophthalazine analogues in order to introduce more nitrogens on the heterocyclic quinones. 1-/2-Substituted-[1,2,3]triazolo[4,5-g]phthalazine-4,9-diones were synthesized by 1,3-dipolar addition of phthalazine-5,8-dione and 4-methoxybenzyl azide by modification of previously reported method. The cytotoxicity of the synthesized compounds was evaluated by a SRB (sulforhodamine B) assay against nine types of human cancer cell lines and inhibition against topoisomerase II (Topo II) of them was assessed by a decatenation assay. Most of the synthesized compounds showed considerably higher cytotoxicity than that of doxorubicin. Also, topoisomerase II inhibitory activity of the tested compounds was higher than that of etoposide and IC(50) values of the compounds were 19.4-64.5 microM.
Sigma-Aldrich
Phthalazine, 98%