Merck
  • Mass analyzed threshold ionization spectroscopy of o-, m-, and p-dichlorobenzenes. Influence of the chlorine position on vibrational spectra and ionization energy.
Mass analyzed threshold ionization spectroscopy of o-, m-, and p-dichlorobenzenes. Influence of the chlorine position on vibrational spectra and ionization energy.
The journal of physical chemistry. A (2008-01-01)
Angela Gaber, Mikko Riese, Juergen Grotemeyer
ABSTRACT
For the first time, vibrational spectra of the 35Cl2 and 35Cl37Cl isotopomers of o-, m-, and p-dichlorobenzene cations in the electronic ground state have been measured via S1 intermediate states by mass analyzed threshold ionization (MATI) spectroscopy. Additionally, ab initio calculations at DFT (density functional theory), CIS (configuration interaction singles), and CASSCF (complete active space self-consistent field) levels of theory have been conducted to compare experimental findings with theory. From the MATI spectra, adiabatic ionization energies of the ortho, meta, and para isomers have been determined to be the same for each pair of investigated isotopomers to 73,237 +/- 6, 72,191 +/- 6, and 73,776 +/- 6 cm(-1), respectively. Several vibrational modes, including fundamentals, combinations, and progressions have been assigned by comparing the experimental and theoretical results. The appearance of overtone progressions involving the 7a mode could be explained by a geometry change of all three isomers during ionization in the direction of this mode by retraining the symmetry of the molecules. Although the general spectral features of the investigated isotopomers are similar, frequencies of some vibrations are slightly different up to a few wavenumbers depending on the involvement of the chlorine atoms in the molecular motion.
Sigma-Aldrich
1,3-Dichlorobenzene, 98%
Sigma-Aldrich
1,3-Dichlorobenzene, puriss., ≥99.0% (GC)