Merck
  • Hexachloroacetone as a precursor for a tetrachloro-substituted oxyallyl intermediate: [4+3] cycloaddition to cyclic 1,3-dienes.
Hexachloroacetone as a precursor for a tetrachloro-substituted oxyallyl intermediate: [4+3] cycloaddition to cyclic 1,3-dienes.
Molecules (Basel, Switzerland) (2007-11-17)
Baldur Föhlisch, Stefan Reiner
ABSTRACT
The enol phosphate 2,2-dichloro-1-(trichloromethyl)ethenyl diethyl phosphate (1), easily available by a Perkow reaction between hexachloroacetone and triethyl phosphite, reacts with sodium trifluoroethoxide/trifluoroethanol in the presence of cyclic 5-membered 1,3-dienes to furnish alpha,alpha,alpha',alpha'-tetrachloro-substituted[3.2.1]bicyclic ketones 2. A [4+3] cycloaddition of a tetrachloro-oxyallyl intermediate is postulated.
Sigma-Aldrich
Hexachloro-2-propanone, 99%
Sigma-Aldrich
Hexachloro-2-propanone, Wacker Chemie AG, ≥99.0% (GC)