Merck
  • Shell cross-linked hyaluronic acid/polylysine layer-by-layer polyelectrolyte microcapsules prepared by removal of reducible hyaluronic acid microgel cores.
Shell cross-linked hyaluronic acid/polylysine layer-by-layer polyelectrolyte microcapsules prepared by removal of reducible hyaluronic acid microgel cores.
Biomacromolecules (2007-11-13)
Hyukjin Lee, Yongho Jeong, Tae Gwan Park
ABSTRACT
Shell cross-linked hollow polyelectrolyte microcapsules composed of hyaluronic acid (HA) and poly- l-lysine (PLL) were prepared by layer-by-layer (LBL) adsorption and subsequent core removal by a reductive agent. Disulfide cross-linked HA microgels were used as template core materials for the LBL deposition on the surface and removed by treatment of dithiothreitol at neutral pH condition. HA/PLL polyelectrolyte multilayers on the shell were chemically cross-linked via carbodiimide chemistry, and their physicochemical properties and drug release behaviors were investigated. Shell cross-linked HA/PLL polyelectrolyte microcapsules exhibited far enhanced physical stability against freeze-thaw cycles and acidic pH conditions compared to the un-cross-linked ones. The cross-linked HA/PLL multilayer shell also demonstrated pH responsive permeability, which became more permeable at low pH than at neutral pH. When bovine serum albumin (BSA), as a model protein drug, was loaded inside using the pH-dependent permeability, BSA release profiles from the microcapsules could be readily modulated by varying medium pH values or adding an HA digesting enzyme (hyaluronidase) in the incubation medium.
Sigma-Aldrich
Poly(isobutylene-alt-maleic anhydride), average Mw ~6,000, 12-200 mesh (85%)
Sigma-Aldrich
Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)], average Mw ~60,000