- Synthesis of alpha-oximinoketones under ultrasound irradiation.
Synthesis of alpha-oximinoketones under ultrasound irradiation.
Ultrasonics sonochemistry (2007-10-26)
Hossein Mehrabi
PMID17959410
ABSTRACT
The reaction of 6-acylmethylphenanthridines with isoamyl nitrite results alpha-oximino-6-acylmethylphenanthridines in 73-95% yields in DMF under ultrasound irradiation. Compared with conventional methods, the main advantages of the present procedure are milder conditions, shorter reaction time and higher yields.