Merck
Effects of the UV filter benzophenone-2 on reproduction in fish.
Toxicology and applied pharmacology (2007-09-25)
Christin J Weisbrod, Petra Y Kunz, Armin K Zenker, Karl Fent
ABSTRACT
The UV filter benzophenone-2 (BP-2) is largely used in personal care products such as cosmetics and in numerous other materials for UV protection. Like other UV filters, BP-2 has been found to be estrogenic in vitro and in vivo, but potential effects on reproduction of fish are unknown. In this study, we evaluate whether BP-2 affects important reproductive parameters such as fecundity, gametogenesis and secondary sex characteristics. After a pre-exposure period of 19 days, reproductively mature fathead minnows (Pimephales promelas) were exposed to 0.002, 0.1, 1.2, 5.0 and 9.7 mg/L BP-2 for 15 days. BP-2 was accumulated in fish up to 3.1 microg/g body weight. In males, a dose-dependent vitellogenin induction and decrease in the number of nuptial tubercles occurred. Moreover, significant dose-related effects on gonads of male and female fish were observed. At concentrations of 1.2 mg/L and higher, spermatocyte and oocyte development was significantly inhibited in male and female fish, respectively. Testes of exposed males had much fewer spermatocytes and ovaries of exposed females had much fewer mature and more atretic follicles. Reproduction was negatively affected in a dose-dependent manner with a decrease in egg production at 5.0 mg/L and a complete cessation of spawning activity at 9.7 mg/L BP-2. Our findings show significant estrogenic effects of the common UV filter BP-2 on vitellogenin induction, secondary sex characteristics, gonadal development, and reproduction in fish.
Sigma-Aldrich
2,2′,4,4′-Tetrahydroxybenzophenone, 97%