Merck
  • Distribution of dimethoate in the body after a fatal organophosphate intoxication.
Distribution of dimethoate in the body after a fatal organophosphate intoxication.
Forensic science international (2007-07-24)
F A Tarbah, A M Shaheen, F A Benomran, A I Hassan, Th Daldrup
ABSTRACT
Many organophosphate pesticides (OPs) such as dimethoate are used to eradicate household pests, and those occurring in agriculture and forestry sectors. Combinations of two or more different insecticides have been manufactured to increase their effectiveness. A case of death is presented as suspected organophosphates intoxication. Autopsy was unremarkable except for grayish fluid in the stomach, with garlicky odor. A systematic toxicological analysis on post-mortem specimens revealed high concentrations of dimethoate in blood 38 microg/mL, urine 0.47 microg/mL, brain 2.2 microg/g, myocardial muscle 7.6 microg/g, liver 4.6 microg/g, lung 7.6 microg/g, skeletal muscle 21 microg/g, kidney 55 microg/g and gall bladder 31 microg/g. Blood alcohol was 2.85 g/L, cyclohexanone and cyclohexanol were also detected in the blood but not quantified. The cause of death was determined as organophosphate intoxication.
Product Description
Supelco
Omethoate, PESTANAL®, analytical standard