- The effect of levocabastine and furosemide pretreatment on hyperreactive response after nasal provocation with hypotonic aerosol in subjects with allergic rhinitis.
The effect of levocabastine and furosemide pretreatment on hyperreactive response after nasal provocation with hypotonic aerosol in subjects with allergic rhinitis.
Patients with allergic rhinitis demonstrate hyperreactive response in distilled water nasal provocation, shown by significant increase in nasal airway resistance (NAR). Antihistamines, including topical antihistamine, levocabastine, reduce response in non-specific nasal provocation tests. Furosemide is a diuretic which reduces hyperreactivity in lower airways, but the mode of its action is not yet fully understood. In this study, we hypothesized that either levocabastine or furosemide pre-treatment in allergic rhinitis patients reduced response to nasal challenge with non-isotonic aerosol. To test the hypothesis, we measured the effect of pre-treatment with levocabastine and furosemide in topical application on suppression of hyperreactive response to distilled water nasal inhalation. Nasal resistance was measured, prior to and after the provocation, by active anterior rhinomanometry in two randomized groups of patients, according to pre-treatment, either by levocabastine or furosemide, 20 patients in each group, respectively. Nasal airflow resistance and level of hyperreactive response considering nasal eosinophilia were tested. Significant increase in nasal resistance following provocation was found at baseline conditions (without pre-medication); pre-treatment with levocabastine and furosemide has suppressed such response. Patients with positive nasal eosinophilia showed a significantly higher increase in nasal resistance compared to those with negative smears. Furosemide has shown significantly better protective effect on nasal resistance increase in patients with positive eosinophils nasal smears. Levocabastine and furosemide pre-treatment suppress hyperreactive response to distilled water nasal provocation. Comparison of resistances (pre-treatment vs. without) showed more protective effect of furosemide, measured on both better and worse patent side of nose, in contrast to levocabastine group for which it was shown only on better patent side prior to provocation. Protection of furosemide was significantly more pronounced in patients with significant nasal eosinophilia.