Merck
  • Origin of a mixed brominated ethene groundwater plume: contaminant degradation pathways and reactions.
Origin of a mixed brominated ethene groundwater plume: contaminant degradation pathways and reactions.
Environmental science & technology (2007-06-28)
Bradley M Patterson, Elizabeth Cohen, Henning Prommer, David G Thomas, Stuart Rhodes, Allan J McKinley
ABSTRACT
On the basis of a combination of laboratory microcosm experiments, column sorption experiments, and the current spatial distribution of groundwater concentrations, the origin of a mixed brominated ethene groundwater plume and its degradation pathway were hypothesized. The contaminant groundwater plume was detected downgradient of a former mineral processing facility, and consisted of tribromoethene (TriBE), cis-1,2-dibromoethene (c-DBE), trans-1,2-dibromoethene (t-DBE), and vinyl bromide (VB). The combined laboratory and field data provided strong evidence that the origin of the mixed brominated ethene plume was a result of dissolution of the dense non-aqueous-phase liquid 1,1,2,2-tetrabromoethane (TBA) atthe presumed source zone, which degraded rapidly (half-life of 0.2 days) to form TriBE in near stoichiometric amounts. TriBE then degraded (half-life of 96 days) to form c-DBE, t-DBE, and VB via a reductive debromination degradation pathway. Slow degradation of c-DBE (half-life >220 days), t-DBE (half-life 220 days), and VB (half-life >220 days) coupled with their low retardation coefficients (1.2, 1.2, and 1.0 respectively) resulted in the formation of an extensive mixed brominated ethene contaminant plume. Without this clearer understanding of the mechanism for TBA degradation, the origin of the mixed brominated ethene groundwater contamination could have been misinterpreted, and inappropriate and ineffective source zone and groundwater remediation techniques could be applied.
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, purum p.a., for separation of mineral compounds, ≥98.0% (GC)