Merck
  • Theophylline granule formulation prepared by the wet granulation method: comparison of in vitro dissolution profiles and estimation of in vivo plasma concentrations.
Theophylline granule formulation prepared by the wet granulation method: comparison of in vitro dissolution profiles and estimation of in vivo plasma concentrations.
European journal of drug metabolism and pharmacokinetics (2007-02-24)
E Karasulu, S Apaydin, I Ince, I Tuglular
ABSTRACT
The primary and secondary objectives of this study were to develop and evaluate the predictability of in vitro-in vivo correlation models for theophylline sustained release (SR) granules. Theophylline SR granules meeting the USP Drug Release Test criteria were prepared using ethyl cellulose (EC) and/or stearyl alcohol (SA) and the wet granulation method. In vitro dissolution studies of granule formulation were performed, and a commercial dosage form was prepared using USP XXIII apparatus II at pH 4.5. Differences and similarities between in vitro dissolution curves were compared using both model-dependent (t-test) and -independent (f1, f2 test) statistical techniques, and it was shown that the three dissolution profiles i.e model-dependent, model-independent, and methods based on ANOVA were very similar. The in vivo performance of the commercial dosage form was tested by oral route using male rabbits and in vitro-in vivo correlations were established. This study indicates that the dosage forms with similar in vitro dissolution profiles may have a similar in vivo performance, and that this performance could be estimated using appropriate correlation equations.
Product Description
Stearyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%
Supelco
1-Octadecanol, Selectophore, ≥99.0%
Sigma-Aldrich
1-Octadecanol, 95%