Merck
  • [Analysis of hydrocarbon impurities in high purity isobutene by gas chromatography].
[Analysis of hydrocarbon impurities in high purity isobutene by gas chromatography].
Se pu = Chinese journal of chromatography (2005-02-17)
Jiwen Li, Yu'an Feng
ABSTRACT
A capillary column HP-AlO2O3/S PLOT(50 m x 0.32 mm i.d. x 8 microm) has been used for the analysis of trace hydrocarbon impurities in high purity isobutene. The results showed that these hydrocarbon impurities could be well separated and with the liquid-phase sample injection valve the disproportionation of sample composition could be avoided. The results also show high accuracy and precision.
Sigma-Aldrich
2-Methylpropene, 99%
Sigma-Aldrich
2-Methylpropene, purum, ≥99.5%