- Effects of redox cycling compounds on glutathione content and activity of glutathione-related enzymes in rainbow trout liver.
Effects of redox cycling compounds on glutathione content and activity of glutathione-related enzymes in rainbow trout liver.
In fish, as in other aerobic organisms, glutathione and glutathione-related enzymes are important components in the defences against oxidative stress. To study if hepatic glutathione levels and/or activities of glutathione-related enzymes can act as indicators of oxidative stress in fish, we injected rainbow trout (Oncorhynchus mykiss) intraperitoneally with paraquat (PQ), menadione (MD), naphthazarin (DHNQ), or beta-naphthoflavone (beta-NF), all known to cause a rise in reactive oxygen species (ROS). After 2 and 5 days of exposure, we measured the activities of hepatic glutathione peroxidase (GPox), glutathione S-transferase (GST), gamma-glutamylcysteine synthetase (GCS), and glutathione reductase (GR). We also measured total glutathione (tGSH) and oxidised glutathione (GSSG) in the liver of fish treated with PQ and MD. All chemicals caused an increase in GR activity after 5 days, which ranged from 160% in fish treated with beta-NF to 1,500% in fish treated with PQ. All chemicals except beta-NF caused moderate elevation in GST activity; GPox activity was lower in fish treated with DHNQ and MD, while GCS activity increased twofold in the fish treated with DHNQ, without being affected by beta-NF, PQ or MD. After 5 days of treatment with PQ or MD, tGSH content was elevated. Our findings demonstrated that of the parameters included in the study, GR activity was the most responsive to treatment with redox cycling compounds, indicating that GR activity is a promising biomarker of such compounds and possibly indicating oxidative stress in rainbow trout.