Merck
  • E-p-methoxycinnamic acid protects cultured neuronal cells against neurotoxicity induced by glutamate.
E-p-methoxycinnamic acid protects cultured neuronal cells against neurotoxicity induced by glutamate.
British journal of pharmacology (2002-03-06)
So Ra Kim, Sang Hyun Sung, Young Pyo Jang, George J Markelonis, Tae H Oh, Young Choong Kim
ABSTRACT
1. We previously reported that four new phenylpropanoid glycosides and six known cinnamate derivatives isolated from roots of Scrophularia buergeriana Miquel (Scrophulariaceae) protected cultured cortical neurons from neurotoxicity induced by glutamate. Here, we have investigated the structure-activity relationships in the phenylpropanoids using our primary culture system. 2. The alpha,beta-unsaturated ester moiety and the para-methoxy group in the phenylpropanoids appeared to play a vital role in neuroprotective activity. This suggested that E-p-methoxycinnamic acid (E-p-MCA) might be a crucial component for their neuroprotective activity within the phenylpropanoid compounds. E-p-MCA significantly attenuated glutamate-induced neurotoxicity when added prior to an excitotoxic glutamate challenge. 3. The neuroprotective activity of E-p-MCA appeared to be more effective in protecting neurons against neurotoxicity induced by NMDA than from that induced by kainic acid. E-p-MCA inhibited the binding of [propyl-2,3-(3)H]-CGP39653 and [2-(3)H]-glycine to their respective binding sites on rat cortical membranes. However, even high concentrations of E-p-MCA failed to inhibit completely [propyl-2,3-(3)H]-CGP39653 and [2-(3)H]-glycine binding. 4. Indeed, E-p-MCA diminished the calcium influx that routinely accompanies glutamate-induced neurotoxicity, and inhibited the subsequent overproduction of nitric oxide and cellular peroxide in glutamate-injured neurons. 5. Thus, our results suggest that E-p-MCA exerts significant protective effects against neurodegeneration induced by glutamate in primary cultures of cortical neurons by an action suggestive of partial glutamatergic antagonism.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%