Merck
  • Investigation of charge localization and charge delocalization in model molecules by multiphoton ionization photoelectron spectroscopy and DFT calculations.
Investigation of charge localization and charge delocalization in model molecules by multiphoton ionization photoelectron spectroscopy and DFT calculations.
Faraday discussions (2000-10-21)
R Weinkauf, F Lehrer, E W Schlag, A Metsala
ABSTRACT
In this work we focus on the question to which degree a surplus charge is localized or delocalized in extended molecular systems. Molecules consisting of a flexible tail and the benzene chromophore, such as n-propylbenzene, 2-phenylethyl alcohol and 2-phenylethylamine, are used as model molecules. Their S0-S1 resonance enhanced multiphoton ionization (MPI) spectra containing origin transitions of different conformers appear at similar wavelengths. This shows, that in the neutral the electronic excitation is localized at the benzene chromophore. Geometry differences between the neutral and the cation can be qualitatively derived from intensities of vibrational transitions or the onset behavior in MPI high-resolution photoelectron (MPI-PE) spectra. We identify two possible reasons for structural changes: Charge-dipole interaction and charge delocalization. Whereas both effects can be active for the folded gauche conformers, the charge-dipole interaction is expected to be small for the extended anti conformers and geometry changes are attributed to charge delocalization. Density functional calculations of structures and energies qualitatively confirm the experimental results for all molecules and their conformers. They predict charge delocalization into the end group of below 20% for n-propylbenzene and 2-phenylethyl alcohol. In the case of 2-phenylethylamine the charge is equally shared by the near-isoenergetic charge sites of the benzene chromophore and the amine group.
Sigma-Aldrich
Propylbenzene, 98%
Sigma-Aldrich
Propylbenzene, ≥99.0% (GC)
Supelco
Propylbenzene, analytical standard