Merck
  • Slow oxidation of acetoxime and methylethyl ketoxime to the corresponding nitronates and hydroxy nitronates by liver microsomes from rats, mice, and humans.
Slow oxidation of acetoxime and methylethyl ketoxime to the corresponding nitronates and hydroxy nitronates by liver microsomes from rats, mice, and humans.
Toxicological sciences : an official journal of the Society of Toxicology (1999-04-30)
W Völkel, N Wolf, M Derelanko, W Dekant
ABSTRACT
Acetoxime and methylethyl ketoxime (MEKO) are tumorigenic in rodents, inducing liver tumors in male animals. The mechanisms of tumorigenicity for these compounds are not well defined. Oxidation of the oximes to nitronates of secondary-nitroalkanes, which are mutagenic and tumorigenic in rodents, has been postulated to play a role in the bioactivation of ketoximes. In these experiments, we have compared the oxidation of acetoxime and methylethyl ketoxime to corresponding nitronates in liver microsomes from different species. The oximes were incubated with liver microsomes from mice, rats, and several human liver samples. After tautomeric equilibration and extraction with n-hexane, 2-nitropropane and 2-nitrobutane were quantitated by GC/MS-NCI (limit of detection of 250 fmol/injection volume). In liver microsomes, nitronate formation from MEKO and acetoxime was dependent on time, enzymatically active proteins, and the presence of NADPH. Nitronate formation was increased in liver microsomes of rats pretreated with inducers of cytochrome P450 and reduced in the presence of inhibitors (n-octylamine and diethyldithiocarbamate). Rates of oxidation of MEKO (Vmax) were 1.1 nmol/min/mg (mice), 0.5 nmol/min/mg (humans), and 0.1 nmol/min/mg (rats). In addition to nitronates, several minor metabolites were also enzymatically formed (two diastereoisomers of 3-nitro-2-butanol, 2-hydroxy-3-butanone oxime and 2-nitro-1-butanol). Acetoxime was also metabolized to the corresponding nitronate at rates approximately 50% of those observed with MEKO oxidation in the three species examined. 2-Nitro-1-propanol was identified as a minor product formed from acetoxime. No sex differences in the capacity to oxidize acetoxime and MEKO were observed in the species examined. The observed results show that formation of sec-nitronates from ketoximes occurs slowly, but is not the only pathway involved in the oxidative biotransformation of these compounds. Due to the lack of sex-specific oxidative metabolism, other metabolic pathways or mechanisms of tumorigenicity not involving bioactivation may be involved in the sex-specific tumorigenicity of ketoximes in rodents.
Sigma-Aldrich
2-Butanone oxime, 99%
Sigma-Aldrich
Acetone oxime, purum, ≥98.0% (GC)
Sigma-Aldrich
3-Nitro-2-butanol, mixture of isomers, 98%
Sigma-Aldrich
Acetone oxime, 98%