Merck
  • Changes in radiation sensitivity and steroid receptor content induced by hormonal agents and ionizing radiation in breast cancer cells in vitro.
Changes in radiation sensitivity and steroid receptor content induced by hormonal agents and ionizing radiation in breast cancer cells in vitro.
Acta oncologica (Stockholm, Sweden) (1996-01-01)
G H Paulsen, T Strickert, A B Marthinsen, S Lundgren
ABSTRACT
Possible influences of tamoxifen and estradiol on in vitro radiation sensitivity and cellular receptor content after irradiation and/or tamoxifen treatment were studied in breast cancer cell lines; estrogen receptor (ER) and progesterone receptor (PgR) positive cell lines MCF-7 and MCF-7/TAM(R)-1 and the ER and PgR negative cell line MDA-MB-231. The tamoxifen resistant MCF-7/TAM(R)-1 cells were more resistant to ionizing radiation than the MCF-7 and MDA-MB-231 cells. Exposure to tamoxifen made the MCF-7 cells more radiation resistant, while estradiol made the MDA-MB-231 cells more radiation sensitive. A radiation dose of 6 Gy reduced the ER content in cytosol in both MCF-7 and MCF-7/TAM(R)-1 cells, but brought no alterations to the PgR content. In MCF-7/TAM(R)-1 cells tamoxifen exposure significantly increased the ER and reduced the PgR content, an effect not observed in the MCF-7 cells. To conclude, the present study indicates that irradiation and tamoxifen may modify the ER and PgR content in cytosol in breast cancer cells. Hormonal treatment may alter the radiation sensitivity, even in ER negative cells, suggesting that hormonal agents may act both via receptor and non-receptor binding mechanisms.
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%