Merck
  • The ubiquitin-specific protease USP25 interacts with three sarcomeric proteins.
The ubiquitin-specific protease USP25 interacts with three sarcomeric proteins.
Cellular and molecular life sciences : CMLS (2006-02-28)
A Bosch-Comas, K Lindsten, R Gonzàlez-Duarte, M G Masucci, G Marfany
ABSTRACT
The biological functions of the more than one hundred genes coding for deubiquitinating enzymes in the human genome remain mostly unknown. The USP25 gene, located at 21q11.2, encodes three protein isoforms produced by alternative splicing. While two of the isoforms are expressed nearly ubiquituously, the expression of the longer USP25 isoform (USP25m) is restricted to muscular tissues and is upregulated during myogenesis. USP25m interacts with three sarcomeric proteins: actin alpha-1 (ACTA1), filamin C (FLNC), and myosin binding protein C1 (MyBPC1), which are critically involved in muscle differentiation and maintenance, and have been implicated in the pathogenesis of severe myopathies. Biochemical analyses demonstrated that MyBPC1 is a short-lived proteasomal substrate, and its degradation is prevented by over-expression of USP25m but not by other USP25 isoforms. In contrast, ACTA1 and FLNC appear to be stable proteins, indicating that their interaction with USP25m is not related to their turnover rate.
Product Description
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein