Merck
  • The complex regulation of human glud1 and glud2 glutamate dehydrogenases and its implications in nerve tissue biology.
The complex regulation of human glud1 and glud2 glutamate dehydrogenases and its implications in nerve tissue biology.
Neurochemistry international (2012-06-05)
Cleanthe Spanaki, Ioannis Zaganas, Zuzana Kounoupa, Andreas Plaitakis
ABSTRACT
Mammalian glutamate dehydrogenase (GDH) is a housekeeping mitochondrial enzyme (hGDH1 in the human) that catalyses the reversible inter-conversion of glutamate to α-ketoglutarate and ammonia, thus interconnecting amino acid and carbohydrate metabolism. It displays an energy sensing mechanism, which permits enzyme activation under low cellular energy states. As GDH is at the crossroads of important metabolic pathways, a tight control of its activity is essential. Indeed, to fulfill its role in metabolism and cellular energetics, mammalian GDH has evolved into a highly regulated enzyme subject to allosteric modulation by diverse compounds. The recent emergence (<23 million years ago) in apes and humans of a hGDH2 isoenzyme with distinct regulatory properties, as well as, the detection of gain-of-function variants in hGDH1 and hGDH2 that affect the nervous system, have introduced additional complexities. The properties of the two highly homologous human GDHs were studied using purified recombinant hGDH1 and hGDH2 obtained by expression of the corresponding cDNAs in Sf21 cells. Results showed that, in contrast to hGDH1 that maintains substantial basal activity (35-40% of its maximal capacity), hGDH2 displays low basal activity (3-8% of maximal) that is remarkably responsive to activation by rising levels of ADP and/or l-leucine. This is primarily due to the Arg443Ser evolutionary change, which also made hGDH2 markedly sensitive to estrogens and neuroleptic drugs. In contrast to hGDH1, which is subject to potent GTP inhibition, hGDH2 has dissociated its function from this energy switch, being able to metabolize glutamate even when the Krebs cycle generates GTP levels sufficient to inactivate the housekeeping hGDH1. Our data also show that spermidine, a polyamine thought to reduce oxidative stress and to prolong survival, and EGCG, a green tea polyphenol, inhibit hGDH2 at lower concentrations than hGDH1. The implications of these findings in nerve tissue biology are discussed.
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type I, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type II, 50% glycerol solution, ≥35 units/mg protein
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type III, lyophilized powder, ≥20 units/mg protein