- Crystal structure analysis of omega-amino acid:pyruvate aminotransferase with a newly developed Weissenberg camera and an imaging plate using synchrotron radiation.
Crystal structure analysis of omega-amino acid:pyruvate aminotransferase with a newly developed Weissenberg camera and an imaging plate using synchrotron radiation.
Journal of biochemistry (1989-01-01)
N Watanabe, K Sakabe, N Sakabe, T Higashi, K Sasaki, S Aibara, Y Morita, K Yonaha, S Toyama, H Fukutani
PMID2500426
ABSTRACT
The three-dimensional structure of omega-amino acid:pyruvate aminotransferase from Pseudomonas sp. F-126, an isologous alpha 4 tetramer containing pyridoxal 5'-phosphate (PLP) as a cofactor, has been determined at 2.0 A resolution. The diffraction data were collected with a newly developed Weissenberg camera with a Fuji Imaging Plate, using synchrotron radiation. The mean figure-of-merit was 0.57. The subunit is rich in secondary structure and comprises two domains. PLP is located in the large domain. The high homology in the secondary structure between this enzyme and aspartate aminotransferase strongly indicates that these two types of enzymes have evolved from a common ancestor.
Sigma-Aldrich
ω-Transaminase, Neosartorya fischeri, recombinant, expressed in E. coli, ≥0.4 U/mg
Sigma-Aldrich
ω-Transaminase, Aspergillus terreus, recombinant, expressed in E. coli, ≥0.10 U/mg
Sigma-Aldrich
ω-Transaminase, Aspergillus fumigatus, recombinant, expressed in E. coli, ≥0.20 U/mg