- Novel synthetic route to the C-nucleoside, 2-deoxy benzamide riboside.
Novel synthetic route to the C-nucleoside, 2-deoxy benzamide riboside.
Bioorganic & medicinal chemistry letters (2012-07-17)
Rebecca R Midtkandal, Philip Redpath, Samuel A J Trammell, Simon J F Macdonald, Charles Brenner, Marie E Migaud
PMID22795628
ABSTRACT
2-Deoxy-C-nucleosides are a subcategory of C-nucleosides that has not been explored extensively, largely because the synthesis is less facile. Flexible synthetic procedures giving access to 2-deoxy-C-nucleosides are therefore of interest. To exemplify the versatility and highlight the limitations of a synthetic route recently developed to that effect, the first synthesis of 2-deoxy benzamide riboside is reported. Biological properties of this novel C-nucleoside are also discussed.
Sigma-Aldrich
Inosine Monophosphate Dehydrogenase Type II human, recombinant, expressed in E. coli