Merck
  • Fast and easy enzyme immobilization by photoinitiated polymerization for efficient bioelectrochemical devices.
Fast and easy enzyme immobilization by photoinitiated polymerization for efficient bioelectrochemical devices.
Analytical chemistry (2011-03-17)
Emmanuel Suraniti, Vincent Studer, Neso Sojic, Nicolas Mano
ABSTRACT
Immobilization and electrical wiring of enzymes is of particular importance for the elaboration of efficient biosensors and can be cumbersome. Here, we report a fast and easy protocol for enzyme immobilization, and as a proof of concept, we applied it to the immobilization of bilirubin oxidase, a labile enzyme. In the first step, bilirubin oxidase is mixed with a redox hydrogel "wiring" the enzyme reaction centers to electrodes. Then, this adduct is covered by an outer layer of PEGDA made by photoinitiated polymerization of poly(ethylene-glycol) diacrylate (PEGDA) and a photoclivable precursor, DAROCUR. This two-step protocol is 18 times faster than the current state-of-the-art protocol and leads to currents 25% higher. In addition, the outer layer of PEGDA acts as a protective layer increasing the lifetime of the electrode by 100% when operating continuously for 2000 s and by 60% when kept in dry state for 24 h. This new protocol is particularly appropriate for labile enzymes that quickly denaturate. In addition, by tuning the ratio PEGDA/DAROCUR, it is possible to make the enzyme electrodes even more active or more stable.
Sigma-Aldrich
Bilirubin Oxidase from Myrothecium verrucaria, lyophilized powder, 15-65 units/mg protein